Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106626.1 | NP_001100096.1 | 5 | 657 | ||
XM_006237595.2 | XP_006237657.1 | 7 | 1373 | |||
XM_008763376.1 | XP_008761598.1 | 7 | 1353 | |||
XM_008763377.1 | XP_008761599.1 | 7 | 1374 |