Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106717.1 | NP_001100187.1 | 8 | 1057 | ||
XM_006239896.1 | XP_006239958.1 | 7 | 884 |