Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750137.3 | XP_003750185.1 | 26 | 4277 | ||
XM_006239996.2 | XP_006240058.1 | 27 | 4146 | |||
XM_006239997.2 | XP_006240059.1 | 27 | 4155 |