Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106733.1 | NP_001100203.1 | 2 | 316 | ||
GenBank mRNA | FQ230024.1 | - | 2 | 79 |