Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282656.1 | NP_001269585.1 | 7, 8 | 766 | ||
NM_001282657.1 | NP_001269586.1 | 6, 7 | 676 | |||
NM_003442.5 | NP_003433.3 | 7, 8 | 769 | |||
XM_005253121.1 | XP_005253178.1 | 7, 8 | 717 | |||
XM_005253122.1 | XP_005253179.1 | 7, 8 | 742 | |||
XM_005253127.1 | XP_005253184.1 | 8, 9 | 795 | |||
XM_011520349.2 | XP_011518651.1 | 7, 8 | 1488 | |||
XM_017018252.1 | XP_016873741.1 | 6, 7 | 653 | |||
XM_017018253.1 | XP_016873742.1 | 6, 7 | 650 | |||
XM_017018254.1 | XP_016873743.1 | 7, 8 | 754 | |||
XM_017018255.1 | XP_016873744.1 | 7, 8 | 844 | |||
XM_017018256.1 | XP_016873745.1 | 6, 7 | 656 | |||
GenBank mRNA | AF071771.1 | - | 4, 5 | 389 | ||
AK291133.1 | - | 7, 8 | 722 | |||
AK297214.1 | - | 6, 7 | 621 | |||
AK313330.1 | - | 7, 8 | 755 | |||
BC020219.1 | - | 7, 8 | 768 | 4902668 |