Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013101.1 | NP_001013119.1 | 2 | 900 | ||
GenBank mRNA | BC091310.1 | - | 2 | 900 | 7311752 |
Gene Symbol: LOC100233176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142942.2 | NP_001136414.1 | 2 | 1020 |