Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106757.2 | NP_001100227.1 | 5 | 647 | ||
GenBank mRNA | BC158889.1 | - | 5 | 647 | 8375684 | |
FQ209917.1 | - | 5 | 678 | |||
FQ212442.1 | - | 5 | 682 |