Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106778.1 | NP_001100248.1 | 21 | 2564 | ||
XM_008765227.1 | XP_008763449.1 | 19 | 7820 | |||
XM_008765228.1 | XP_008763450.1 | 19 | 1855 | |||
XM_008765229.1 | XP_008763451.1 | 18 | 7775 | |||
XM_008765230.1 | XP_008763452.1 | 18 | 1750 | |||
XM_008765231.1 | XP_008763453.1 | 23 | 3488 | |||
GenBank mRNA | FQ213292.1 | - | 2 | 131 |