Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765227.1 | XP_008763449.1 | 6, 7 | 6502 | ||
XM_008765228.1 | XP_008763450.1 | 6, 7 | 537 | |||
XM_008765229.1 | XP_008763451.1 | 5, 6 | 6457 | |||
XM_008765230.1 | XP_008763452.1 | 5, 6 | 432 | |||
XM_008765231.1 | XP_008763453.1 | 10, 11 | 2170 |