Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF113A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006978.2 | NP_008909.1 | 1 | 411 | ||
GenBank mRNA | AK314778.1 | - | 1 | 357 | ||
AW001915.1 | - | 1 | 411 | |||
BC000832.1 | - | 1 | 254 | 3455861 | ||
BC020556.1 | - | 1 | 297 | 3447759 | ||
BC128440.1 | - | 1 | 296 | |||
BT007175.1 | - | 1 | 196 | |||
X98253.1 | - | 1 | 406 |