Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 5 | 1006 | ||
NM_001317387.1 | NP_001304316.1 | 4 | 925 | |||
NM_001650.5 | NP_001641.1 | 5 | 1006 | |||
NM_004028.4 | NP_004019.1 | 4 | 1133 | |||
XM_011525942.2 | XP_011524244.1 | 5 | 1048 | |||
GenBank mRNA | AB128929.1 | - | 6 | 536 | ||
AB209156.1 | - | 3 | 539 | |||
AK222684.1 | - | 5 | 918 | |||
AK295069.1 | - | 6 | 817 | |||
BC022286.1 | - | 5 | 871 | 4717755 | ||
D63412.1 | - | 5 | 871 | |||
HQ901095.1 | - | 5 | 980 | |||
KF055862.1 | - | 4 | 770 | |||
U63622.1 | - | 5 | 890 | |||
U63623.1 | - | 4 | 794 |