Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765827.1 | XP_008764049.1 | 17, 18 | 2577 | ||
XM_008765828.1 | XP_008764050.1 | 17, 18 | 2577 | |||
XM_008765829.1 | XP_008764051.1 | 16, 17 | 2538 | |||
XM_008765830.1 | XP_008764052.1 | 16, 17 | 2538 | |||
XM_008765831.1 | XP_008764053.1 | 16, 17 | 2538 | |||
XM_008765832.1 | XP_008764054.1 | 16, 17 | 2538 | |||
XM_008765833.1 | XP_008764055.1 | 16, 17 | 2538 | |||
XM_008765834.1 | XP_008764056.1 | 16, 17 | 2538 | |||
XM_008765835.1 | XP_008764057.1 | 16, 17 | 2538 | |||
XM_008765836.1 | XP_008764058.1 | 16, 17 | 2538 |