Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or10ad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765883.1 | XP_008764105.1 | 1 | 664 |
Gene Symbol: Olr1102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765851.1 | XP_008764073.1 | 2 | 656 |