Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013249.2 | NP_037381.2 | 3 | 1122 | ||
XM_005253128.3 | XP_005253185.1 | 4 | 1197 | |||
XM_005253129.3 | XP_005253186.1 | 3 | 1130 | |||
XM_006718308.3 | XP_006718371.1 | 3 | 1130 | |||
XM_006718309.3 | XP_006718372.1 | 3 | 1126 | |||
XM_011520355.2 | XP_011518657.1 | 4 | 1193 | |||
XM_011520357.2 | XP_011518659.1 | 3 | 884 | |||
XM_017018269.1 | XP_016873758.1 | 3 | 898 | |||
GenBank mRNA | AF056617.1 | - | 3 | 1077 | ||
AK313462.1 | - | 3 | 1094 | |||
BC136772.1 | - | 3 | 1002 | 9020762 | ||
BC144405.1 | - | 4 | 1061 | 9052926 |