Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106798.2 | NP_001100268.2 | 3 | 524 | ||
GenBank mRNA | BC098729.1 | - | 2 | 880 | ||
BC166800.1 | - | 3 | 524 | 7938259 | ||
FQ209549.1 | - | 3 | 540 | |||
FQ214028.1 | - | 3 | 543 | |||
FQ214274.1 | - | 3 | 537 | |||
FQ214451.1 | - | 3 | 538 | |||
FQ220704.1 | - | 3 | 538 | |||
FQ230080.1 | - | 3 | 543 |