Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013954.1 | NP_001013976.1 | 9 | 2429 | ||
XM_008766249.1 | XP_008764471.1 | 8 | 2373 | |||
XM_008766250.1 | XP_008764472.1 | 7 | 2254 | |||
XM_008766251.1 | XP_008764473.1 | 9 | 2464 | |||
XM_008766252.1 | XP_008764474.1 | 8 | 2317 | |||
XM_008766253.1 | XP_008764475.1 | 8 | 2760 | |||
XM_008766255.1 | XP_008764477.1 | 9 | 2489 | |||
XM_008766256.1 | XP_008764478.1 | 8 | 2295 | |||
XM_008766257.1 | XP_008764479.1 | 8 | 2934 | |||
XM_008766258.1 | XP_008764480.1 | 8 | 5958 | |||
GenBank mRNA | BC083657.1 | - | 7 | 1508 | ||
BC085805.1 | - | 9 | 2429 | 7112021 |