Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 19 | 2827 | ||
NM_001130523.2 | NP_001123995.1 | 19 | 2881 | |||
NM_001242891.1 | NP_001229820.1 | 20 | 2974 | |||
NM_001242892.1 | NP_001229821.1 | 18 | 2440 | |||
NM_001242893.1 | NP_001229822.1 | 17 | 2347 | |||
NM_007158.5 | NP_009089.4 | 18 | 2734 | |||
GenBank mRNA | AB020692.1 | - | 19 | 2741 | ||
AF077054.1 | - | 18 | 2650 | |||
AK022516.1 | - | 17 | 2249 | |||
AK022528.1 | - | 17 | 2249 | |||
AK130392.1 | - | 7 | 935 | |||
AK290146.1 | - | 18 | 2372 | |||
AL050150.1 | - | 8 | 1027 | |||
AL110192.1 | - | 3 | 433 | |||
AY049788.1 | - | 16 | 2237 | |||
BC021692.1 | - | 6 | 737 | |||
BC032446.1 | - | 18 | 2669 | 5111213 | ||
BX648561.1 | - | 18 | 2571 | |||
BX648780.1 | - | 18 | 2668 | |||
CR749378.1 | - | 19 | 2831 | |||
CR749431.1 | - | 19 | 2813 |