Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767310.1 | XP_008765532.1 | 8251 | |||
XM_008767311.1 | XP_008765533.1 | 8251 | ||||
XM_008767312.1 | XP_008765534.1 | 8224 | ||||
XM_008767313.1 | XP_008765535.1 | 8251 | ||||
XM_008767314.1 | XP_008765536.1 | 8251 | ||||
XM_008767315.1 | XP_008765537.1 | 8251 |