Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100672.1 | NP_001094142.1 | 3 | 308 | ||
XM_008773408.1 | XP_008771630.1 | 4 | 568 | |||
GenBank mRNA | BC086430.1 | - | 2 | 148 | ||
BC158651.1 | - | 3 | 308 | 8368067 | ||
FQ213424.1 | - | 3 | 365 | |||
FQ216491.1 | - | 3 | 331 |