Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256992.2 | XP_006257054.1 | 2 | 889 | ||
XM_008773286.1 | XP_008771508.1 | 2 | 844 | |||
XM_008773287.1 | XP_008771509.1 | 1 | 757 | |||
XM_008773288.1 | XP_008771510.1 | 2 | 815 |