Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127532.1 | NP_001121004.1 | 4 | 774 | ||
XM_008767481.1 | XP_008765703.1 | 3 | 980 | |||
GenBank mRNA | BC166713.1 | - | 4 | 774 | 7930724 |