Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106978.1 | NP_001100448.1 | 7 | 1200 | ||
XM_006245801.2 | XP_006245863.1 | 7 | 1247 | |||
XM_008767487.1 | XP_008765709.1 | 6 | 1322 | |||
XM_008767488.1 | XP_008765710.1 | 6 | 1319 |