Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106982.1 | NP_001100452.1 | 6 | 1028 | ||
GenBank mRNA | BC161878.1 | - | 6 | 976 | 7123846 |