Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727761.4 | XP_002727807.1 | 14 | 2411 | ||
XM_008767624.1 | XP_008765846.1 | 14 | 2806 | |||
XM_008767625.1 | XP_008765847.1 | 14 | 2523 | |||
GenBank mRNA | FQ212171.1 | - | 11 | 1527 |