Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007670.1 | NP_001007671.1 | 4 | 712 | ||
XM_006246698.2 | XP_006246760.1 | 4 | 1134 | |||
GenBank mRNA | BC079319.1 | - | 4 | 712 | 7135137 |