Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191820.1 | NP_001178749.1 | 32, 33 | 4876 | ||
XM_008768006.1 | XP_008766228.1 | 32, 33 | 4537 | |||
XM_008768007.1 | XP_008766229.1 | 33, 34 | 4728 | |||
XM_008768008.1 | XP_008766230.1 | 32, 33 | 4728 | |||
GenBank mRNA | BC098834.1 | - | 1, 2 | 135 | ||
BC169093.1 | - | 3, 4 | 452 |