Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039018.1 | NP_001034107.1 | 3 | 1339 | ||
XM_006246727.2 | XP_006246789.1 | 4 | 1596 | |||
XM_006246729.2 | XP_006246791.1 | 4 | 1436 | |||
GenBank mRNA | BC082089.1 | - | 4 | 1389 | ||
BC100079.1 | - | 3 | 1339 | 7132945 |