Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107040.1 | NP_001100510.1 | 2, 3 | 472 | ||
GenBank mRNA | HQ616894.1 | - | 2, 3 | 494 |