Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012237.1 | NP_001012237.1 | 6 | 887 | ||
XM_006247610.2 | XP_006247672.1 | 6 | 989 |