Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013975.2 | NP_001013997.2 | 8 | 942 | ||
XM_006247887.2 | XP_006247949.1 | 8 | 1175 | |||
XM_006247888.2 | XP_006247950.1 | 7 | 1512 | |||
GenBank mRNA | BC090016.1 | - | 1 | 324 | ||
CO562343.1 | - | 3 | 275 |