Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myadml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128185.1 | NP_001121657.1 | 1 | 512 | ||
GenBank mRNA | BC167064.1 | - | 1 | 512 | 7193100 |