Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195626.1 | NP_001182555.1 | 17 | 2313 | ||
NM_001324297.1 | NP_001311226.1 | 20 | 3146 | |||
NM_004641.3 | NP_004632.1 | 19 | 2710 | |||
NR_136736.1 | 21 | 2958 | ||||
GenBank mRNA | AB209755.1 | - | 21 | 2664 | ||
AY598745.1 | - | 17 | 2313 | |||
BC129946.1 | - | 18 | 2356 | |||
BX640774.1 | - | 7 | 911 | |||
BX648210.1 | - | 11 | 1398 | |||
U13948.1 | - | 19 | 2544 |