Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079938.1 | NP_001073407.1 | 3 | 512 | ||
XM_008768634.1 | XP_008766856.1 | 3 | 404 | |||
XM_008768635.1 | XP_008766857.1 | 3 | 599 | |||
XM_008768636.1 | XP_008766858.1 | 4 | 713 | |||
XM_008768637.1 | XP_008766859.1 | 4 | 518 | |||
XM_008768638.1 | XP_008766860.1 | 3 | 404 | |||
XM_008768639.1 | XP_008766861.1 | 3 | 599 | |||
XM_008768640.1 | XP_008766862.1 | 3 | 290 | |||
XM_008768641.1 | XP_008766863.1 | 3 | 404 | |||
GenBank mRNA | BC129113.1 | - | 3 | 512 | 7112122 |