Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc8e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034139.1 | NP_001029311.1 | 3 | 1201 | ||
XM_006248763.2 | XP_006248825.1 | 3 | 1237 | |||
XM_006248764.2 | XP_006248826.1 | 4 | 1370 | |||
XM_008768954.1 | XP_008767176.1 | 3 | 1505 | |||
XM_008768955.1 | XP_008767177.1 | 2 | 1034 | |||
GenBank mRNA | BC105779.1 | - | 3 | 1201 | 7932319 |