Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pds5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101805.1 | NP_001095275.1 | 3 | 383 | ||
NM_001102383.2 | NP_001095853.1 | 3 | 383 | |||
XM_008768957.1 | XP_008767179.1 | 4 | 469 | |||
XM_008768958.1 | XP_008767180.1 | 4 | 500 | |||
XM_008768959.1 | XP_008767181.1 | 4 | 412 | |||
GenBank mRNA | AY388627.1 | - | 2 | 253 |