Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013438.2 | NP_001013456.1 | 14 | 1499 | ||
NM_001013439.2 | NP_001013457.1 | 15 | 1843 | |||
NM_005087.3 | NP_005078.2 | 14 | 1499 | |||
XM_005247813.3 | XP_005247870.2 | 13 | 1361 | |||
XM_005247814.3 | XP_005247871.2 | 13 | 1361 | |||
XM_005247815.3 | XP_005247872.2 | 14 | 1274 | |||
XM_005247816.3 | XP_005247873.2 | 13 | 1361 | |||
XM_006713775.3 | XP_006713838.1 | 14 | 1704 | |||
XM_017007298.1 | XP_016862787.1 | 14 | 1704 | |||
XM_017007299.1 | XP_016862788.1 | 15 | 1617 | |||
XM_017007300.1 | XP_016862789.1 | 14 | 1704 | |||
XM_017007301.1 | XP_016862790.1 | 15 | 1617 | |||
GenBank mRNA | AK225334.1 | - | 14 | 1275 | ||
AK292633.1 | - | 14 | 1275 | |||
AK294603.1 | - | 13 | 1221 | |||
AK297254.1 | - | 12 | 1127 | |||
AK302197.1 | - | 14 | 1278 | |||
AY341428.1 | - | 14 | 1550 | |||
BC028983.1 | - | 14 | 1269 | 5122574 | ||
KR259877.1 | - | 14 | 1259 | |||
KR259878.1 | - | 13 | 1346 | |||
KR259879.1 | - | 13 | 1346 | |||
KR259880.1 | - | 15 | 1571 | |||
U25165.1 | - | 14 | 1271 |