Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107195.1 | NP_001100665.1 | 7 | 2018 | ||
XM_006250219.2 | XP_006250281.1 | 7 | 2039 |