Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013127.1 | NP_001013145.1 | 5 | 550 | ||
XM_006250295.2 | XP_006250357.1 | 5 | 985 | |||
GenBank mRNA | BC084703.1 | - | 5 | 550 | 7300611 | |
FQ221577.1 | - | 5 | 561 | |||
FQ223112.1 | - | 5 | 575 | |||
FQ228757.1 | - | 5 | 569 |