Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025670.1 | NP_001020841.1 | 5 | 932 | ||
XM_008770019.1 | XP_008768241.1 | 6 | 929 | |||
XM_008770020.1 | XP_008768242.1 | 6 | 1358 | |||
XM_008770021.1 | XP_008768243.1 | 6 | 1057 | |||
GenBank mRNA | BC097362.1 | - | 5 | 932 | 7442656 |