Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100564.1 | NP_001094034.1 | 7, 8 | 1557 | ||
XM_006250657.2 | XP_006250719.1 | 8, 9 | 1673 | |||
XM_006250658.2 | XP_006250720.1 | 8, 9 | 1703 | |||
XM_006250659.2 | XP_006250721.1 | 8, 9 | 1868 | |||
XM_006250661.2 | XP_006250723.1 | 8, 9 | 1040 | |||
XM_008770022.1 | XP_008768244.1 | 8, 9 | 1010 | |||
GenBank mRNA | BC082043.1 | - | 5, 6 | 763 | ||
BC099076.1 | - | 5, 6 | 594 |