Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013430.1 | NP_001013448.1 | 3 | 834 | ||
GenBank mRNA | BC091269.1 | - | 3 | 834 | 7381512 | |
FQ225173.1 | - | 3 | 846 | |||
FQ228192.1 | - | 3 | 922 | |||
FQ232983.1 | - | 3 | 846 |