Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770170.1 | XP_008768392.1 | 9 | 2122 | ||
XM_008770171.1 | XP_008768393.1 | 10 | 2320 | |||
XM_008770172.1 | XP_008768394.1 | 7 | 2855 | |||
XM_008770173.1 | XP_008768395.1 | 8 | 1902 | |||
XM_008770174.1 | XP_008768396.1 | 8 | 1786 | |||
XM_008770175.1 | XP_008768397.1 | 7 | 1699 |