Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006532.3 | NP_006523.1 | 6 | 866 | ||
XM_011528330.2 | XP_011526632.2 | 6 | 868 | |||
XM_017027335.1 | XP_016882824.1 | 7 | 850 | |||
XM_017027336.1 | XP_016882825.1 | 2 | 602 | |||
GenBank mRNA | AB208794.1 | - | 7 | 847 | ||
AK225481.1 | - | 6 | 805 | |||
AK293105.1 | - | 6 | 799 | |||
BC049195.1 | - | 6 | 848 | 5170024 | ||
U16282.1 | - | 6 | 806 |