Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130062.1 | NP_001123534.1 | 3 | 300 | ||
XM_006251239.2 | XP_006251301.1 | 4 | 1117 | |||
XM_008770283.1 | XP_008768505.1 | 4 | 1117 | |||
GenBank mRNA | FQ223777.1 | - | 4 | 419 |