Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191683.1 | NP_001178612.1 | 2 | 443 | ||
XM_006251579.2 | XP_006251641.2 | 2 | 229 | |||
XM_006251586.2 | XP_006251648.1 | 2 | 666 | |||
XM_006251587.2 | XP_006251649.1 | 2 | 364 | |||
XM_006251588.2 | XP_006251650.1 | 2 | 665 | |||
XM_006251594.2 | XP_006251656.1 | 2 | 652 | |||
XM_008770430.1 | XP_008768652.1 | 2 | 665 | |||
XM_008770431.1 | XP_008768653.1 | 2 | 667 | |||
GenBank mRNA | BC086607.1 | - | 2 | 552 | ||
BC158689.1 | - | 2 | 356 | 8375842 |