Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723901.3 | XP_006723964.1 | 4 | 524 | ||
XM_006723902.3 | XP_006723965.1 | 4 | 513 | |||
XM_006723903.3 | XP_006723966.1 | 4 | 511 | |||
XM_006723904.3 | XP_006723967.1 | 4 | 512 | |||
XM_011546081.2 | XP_011544383.1 | 3 | 395 | |||
XM_017028514.1 | XP_016884003.1 | 4 | 514 | |||
XM_017028516.1 | XP_016884005.1 | 3 | 677 | |||
XM_017028517.1 | XP_016884006.1 | 3 | 312 |
Gene Symbol: C21orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320383.1 | NP_001307312.1 | 4 | 501 | ||
NM_001320384.1 | NP_001307313.1 | 4 | 501 | |||
NM_004649.7 | NP_004640.4 | 4 | 501 | |||
NM_198155.4 | NP_937798.4 | 4 | 501 | |||
NR_135220.1 | 3 | 448 | ||||
XM_017028479.1 | XP_016883968.1 | 3 | 312 | |||
GenBank mRNA | BC002370.2 | - | 4 | 486 | 2822340 | |
BC003587.1 | - | 4 | 432 | 2822714 | ||
BI196781.1 | - | 4 | 376 | |||
BP283346.1 | - | 4 | 501 | |||
BQ647697.1 | - | 4 | 334 | |||
BQ882034.1 | - | 3 | 305 | |||
CX788692.1 | - | 4 | 493 | |||
D86061.1 | - | 4 | 432 | |||
D86062.1 | - | 4 | 433 | |||
U53003.1 | - | 4 | 499 | |||
Y07572.1 | - | 4 | 482 |