Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107284.1 | NP_001100754.1 | 1 | 604 | ||
XM_006252444.2 | XP_006252506.1 | 2 | 1209 | |||
XM_006252445.2 | XP_006252507.1 | 2 | 872 | |||
XM_006252446.2 | XP_006252508.1 | 2 | 682 | |||
XM_006252447.2 | XP_006252509.1 | 2 | 978 |