Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duxbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252678.2 | XP_006252740.1 | 6 | 3718 | ||
XM_006252679.2 | XP_006252741.1 | 7 | 1719 | |||
XM_008771030.1 | XP_008769252.1 | 4 | 1210 | |||
XM_008771031.1 | XP_008769253.1 | 5 | 4393 | |||
XM_224643.6 | XP_224643.1 | 6 | 2394 |