Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252707.2 | XP_006252769.1 | 1 | 466 | ||
XM_006252708.2 | XP_006252770.1 | 1 | 466 | |||
XM_008771058.1 | XP_008769280.1 | 3 | 6274 |