Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004651.3 | NP_004642.2 | 13 | 1834 | ||
XM_005272674.3 | XP_005272731.1 | 13 | 1671 | |||
XM_011543988.1 | XP_011542290.1 | 13 | 1628 | |||
GenBank mRNA | AB073597.1 | - | 13 | 1736 | ||
AK055588.1 | - | 12 | 1891 | |||
AK098394.1 | - | 13 | 1738 | |||
AK294635.1 | - | 13 | 1580 | |||
BC000350.2 | - | 13 | 1708 | |||
BC063668.1 | - | 13 | 1716 | |||
BC140849.1 | - | 13 | 1883 | 9021717 | ||
U44839.1 | - | 13 | 1694 |